3-Chloro-4-fluoroaniline 98

Supplier: Thermo Fisher Scientific
Danger

A11858.14 A11858.22 A11858.36
A11858.14EA 29.4 EUR
A11858.14 A11858.22 A11858.36
3-Chloro-4-fluoroaniline 98
3-Chloro-4-fluoroaniline

Purity: 98+% CAS No.: 367-21-5 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Flash Point: 149°(300°F) Odour: Characteristic Boiling Point: 227-230° Beilstein Reference: 1562786 UN No.: 2811 MDL No.: MFCD00007767

Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 227…230 °C
Melting Pt: 44…47 °C
Density: 1.420
Flash Pt: 149 °C (300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR