4-Ethylmorpholine ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Ethylmorpholine

A11905.AE A11905.AK A11905.AP A11905.AU
A11905.AEEA 26.1 EUR
A11905.AE A11905.AK A11905.AP A11905.AU
4-Ethylmorpholine ≥98%
4-Ethylmorpholine

Purity: 98% CAS No.: 100-74-3 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 27°(80°F) Refractive Index: 1.4413 Melting Point: -63° Boiling Point: 138-139° Beilstein Reference: 102969 UN No.: 2734 MDL No.: MFCD00006177

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 138…139 °C
Melting Pt: –63 °C
Density: 0.908
Flash Pt: 27 °C (80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR