L(+)-Ornithine hydrochloride ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

A12111.14 A12111.22 A12111.36
A12111.14EA 31.7 EUR
A12111.14 A12111.22 A12111.36
L(+)-Ornithine hydrochloride ≥99%
L(+)-Ornithine hydrochloride

Purity: 99% CAS No.: 3184-13-2 Molecular Formula: C5H12N2O2.HCl Formula Weight: 168.62 Melting Point: ca 238° dec. Beilstein Reference: 3625847 Merck Reference: 14,6874 Optical Rotation: +23° (c=5 in 5N HCl) MDL No.: MFCD00064562

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Melting Pt: ∼ 238 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR