2-Fluorobiphenyl 98

Supplier: Thermo Fisher Scientific

Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1

A12227.06 A12227.09 A12227.14
A12227.06EA 80.6 EUR
A12227.06 A12227.09 A12227.14
2-Fluorobiphenyl 98
2-Fluorobiphenyl

Purity: 98% CAS No.: 321-60-8 Molecular Formula: C12H9F Formula Weight: 172.20 Melting Point: 72-76° Boiling Point: 248° MDL No.: MFCD00000317

Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 248 °C
Melting Pt: 72…76 °C
Density: 1.245
Storage Temperature: Ambient
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR