4-Fluorophenyl isocyanate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

A12576.03 A12576.06 A12576.14
A12576.03EA 28 EUR
A12576.03 A12576.06 A12576.14
4-Fluorophenyl isocyanate ≥98%
4-Fluorophenyl isocyanate

Purity: 98+% CAS No.: 1195-45-5 Molecular Formula: C7H4FNO Formula Weight: 137.11 Storage Temperature: 2°C to 8°C Flash Point: 52°(125°F) Refractive Index: 1.5140 Odour: Pungent, makes eyes water Boiling Point: 55-56°/8mm Beilstein Reference: 471609 UN No.: 3080 MDL No.: MFCD00002023

Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 55…56 °C (8 mmHg)
Density: 1.210
Flash Pt: 52 °C (125 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS Number: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR