1-Butanesulphonic acid sodium salt 99

Supplier: Thermo Fisher Scientific

Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt

A12654.06 A12654.14 A12654.22
A12654.06EA 53.4 EUR
A12654.06 A12654.14 A12654.22
1-Butanesulphonic acid sodium salt 99
1-Butansulphonic acid sodium salt

Purity: 99% CAS No.: 2386-54-1 Molecular Formula: CH3(CH2)3SO3Na Formula Weight: 160.17 Melting Point: 300° Beilstein Reference: 3716662

Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR