3-Methoxyphenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Hydroxy-3-methoxybenzene, m-Methoxyphenol

A12826.14 A12826.22 A12826.36
A12826.14EA 28.9 EUR
A12826.14 A12826.22 A12826.36
3-Methoxyphenol ≥97%
3-Methoxyphenol

Purity: 97% CAS No.: 150-19-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 124°(255°F) Refractive Index: 1.5520 Odour: Sweet Melting Point: -18° Boiling Point: 244° Beilstein Reference: 1209898 UN No.: 2810 MDL No.: MFCD00002267

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 244 °C
Melting Pt: –18 °C
Density: 1.140
Flash Pt: 124 °C (255 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR