2,5-Dimethoxybenzoic acid 98

Supplier: Thermo Fisher Scientific
Warning

A12901.06 A12901.14 A12901.22
A12901.06EA 67.1 EUR
A12901.06 A12901.14 A12901.22
2,5-Dimethoxybenzoic acid 98
2,5-Dimethoxybenzoic acid

Purity: 98+% CAS No.: 2785-98-0 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odour: Odourless Melting Point: 75-79° Beilstein Reference: 2097993 MDL No.: MFCD00002436

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 314
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002436
CAS Number: 2785-98-0
EINECS: 220-503-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR