Thioacetamide 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethanethioamide

A12926.14 A12926.22 A12926.36
A12926.14EA 35.5 EUR
A12926.14 A12926.22 A12926.36
Thioacetamide 98
Thioacetamide

Purity: 98% CAS No.: 62-55-5 Molecular Formula: C2H5NS Formula Weight: 75.13 Melting Point: 110-114° Solubility: Soluble in water, ethanol Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 110…114 °C
Density: 1.37
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR