Oil red O

Supplier: Thermo Fisher Scientific

Synonyms: CI 26125, Sudan Red 5B, Solvent Red 27

A12989.14 A12989.22 A12989.A1
A12989.14EA 31.3 EUR
A12989.14 A12989.22 A12989.A1
Oil red O
Oil Red O

CAS No.: 1320-06-5 Molecular Formula: C26H24N4O Formula Weight: 408.51 Melting Point: ca 166° dec. Beilstein Reference: 3491382 MDL No.: MFCD00003898

Formula: C₂₆H₂₄N₄O
MW: 408.51 g/mol
Melting Pt: ∼ 166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003898
CAS Number: 1320-06-5
EINECS: 215-295-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR