2-Bromo-m-xylene 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromo-1,3-dimethylbenzene, 2,6-Dimethylbromobenzene

A13130.09 A13130.14 A13130.22 A13130.30 A13130.36
A13130.09EA 39.8 EUR
A13130.09 A13130.14 A13130.22 A13130.30 A13130.36
2-Bromo-m-xylene 98
2-Bromo-m-xylene

Purity: 98+% CAS No.: 576-22-7 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 73°(163°F) Refractive Index: 1.5550 Melting Point: -10° Boiling Point: 205-206° Beilstein Reference: 1929780 MDL No.: MFCD00000075

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 205…206 °C
Melting Pt: –10 °C
Density: 1.384
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000075
CAS Number: 576-22-7
EINECS: 209-397-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR