3-Amino-4-pyrazolecarbonitrile 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Aminopyrazole-4-carbonitrile, 3-Amino-1H-pyrazole-4-carbonitrile, 3-Aminopyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole 3-Amino-4-pyrazolecarbonitrile 5-Amino-4-cyanopyrazole 3-Amino-1H-pyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole

A14266.06 A14266.14
A14266.06EA 81.5 EUR
A14266.06 A14266.14
3-Amino-4-pyrazolecarbonitrile 98%
3-Amino-4-pyrazolecarbonitrile

Purity: 98% CAS No.: 16617-46-2 Molecular Formula: C4H4N4 Formula Weight: 108.10 Odour: Odourless Melting Point: 173-176° Beilstein Reference: 2647 UN No.: UN3439 MDL No.: MFCD00005237

Formula: C₄H₄N₄
MW: 108.10 g/mol
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00005237
CAS Number: 16617-46-2
EINECS: 240-665-6
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR