4-Chlorophenylhydrazinium chloride ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride

A14518.06 A14518.14 A14518.22
A14518.06EA 32.2 EUR
A14518.06 A14518.14 A14518.22
4-Chlorophenylhydrazinium chloride ≥97%
4-Chlorophenylhydrazinium chloride

Purity: 97% CAS No.: 1073-70-7 Molecular Formula: C6H7ClN2.HCl Formula Weight: 179.05 Odour: Odourless Melting Point: ca 216° dec. Beilstein Reference: 3596254 MDL No.: MFCD00012943

Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
Melting Pt: ∼ 216 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR