L-Arginine hydrochloride 98

Supplier: Thermo Fisher Scientific

Synonyms: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride

A14730.0E A14730.22 A14730.36
A14730.0EEA 578 EUR
A14730.0E A14730.22 A14730.36
L-Arginine hydrochloride 98
L-Arginine hydrochloride

Purity: 98+% CAS No.: 1119-34-2 Molecular Formula: C6H14N4O2.HCl Formula Weight: 210.67 Melting Point: 216° dec. Beilstein Reference: 3631658 Merck Reference: 14,780 Optical Rotation: +22° (c=5 in 5N HCl) MDL No.: MFCD00064550

Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 216 °C (dec.)
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR