Potassium tetraoxalate dihydrate 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate

A14933.30 A14933.0I A14933.0B
A14933.30EA 39.8 EUR
A14933.30 A14933.0I A14933.0B
Potassium tetraoxalate dihydrate 98
Potassium tetraoxalate dihydrate

Purity: 98+% CAS No.: 6100-20-5 Molecular Formula: KH3(C2O4)2.2H2O Formula Weight: 254.19 (218.16anhy) Merck Reference: 14,7689 UN No.: 3288 MDL No.: MFCD00150443

Formula: C₄H₃KO₈·2H₂O
MW: 254.19 g/mol (218.16 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150443
CAS Number: 6100-20-5
EINECS: 204-874-6
UN: 3288
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR