Magnesium stearate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt

A16244.0B A16244.30
A16244.0BEA 84.5 EUR
A16244.0B A16244.30
Magnesium stearate
Magnesium stearate

CAS No.: 557-04-0 Molecular Formula: [CH3(CH2)16CO2]2Mg Formula Weight: 591.27 Physical Form: Powder Odour: Odourless Melting Point: 148-152° MDL No.: MFCD00036391

Formula: C₃₆H₇₀MgO₄
MW: 591.25 g/mol
Melting Pt: 148…152 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR