1-Chloro-3-methyl-2-butene ≥95% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilization: Stabilised with potassium carbonate

A16545.18
A16545.18EA 304 EUR
A16545.18
1-Chloro-3-methyl-2-butene ≥95% stabilised
1-Chloro-3-methyl-2-butene

Stabilized with potassium carbonate


Purity: 95% CAS No.: 503-60-6 Molecular Formula: C5H9Cl Formula Weight: 104.57 Storage Temperature: 2°C to 8°C Flash Point: 13°(56°F) Refractive Index: 1.4490 Boiling Point: 107-109° UN No.: 1993 MDL No.: MFCD00000982

Formula: C₅H₉Cl
MW: 104.58 g/mol
Boiling Pt: 107…109 °C
Density: 0.928
Flash Pt: 13 °C (56 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000982
CAS Number: 503-60-6
EINECS: 207-972-7
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR