L(+)-Lactic acid ∼85 - 90% aqueous solution

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

A17268.0E A17268.22 A17268.36 AB117182.0100
A17268.0EEA 230 EUR
A17268.0E A17268.22 A17268.36 ABCRAB117182.0100
L(+)-Lactic acid ∼85 - 90% aqueous solution
L(+)-Lactic acid

CAS No.: 79-33-4 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 110°(230°F) Refractive Index: 1.4270 Odour: Odourless pH: 2.8 Melting Point: 18° Beilstein Reference: 1720251 Merck Reference: 14,5337 Optical Rotation: -13° (c=2.5 in 1N NaOH) MDL No.: MFCD00064266

Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: 18 °C
Density: 1.209
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 14,5337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR