2-Fluorophenyl isocyanate 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Fluoro-2-isocyanatobenzene

A17466.03 A17466.06 A17466.14
A17466.03EA 27.6 EUR
A17466.03 A17466.06 A17466.14
2-Fluorophenyl isocyanate 98
2-Fluorophenyl isocyanate

Purity: 98% CAS No.: 16744-98-2 Molecular Formula: C7H4FNO Formula Weight: 137.11 Storage Temperature: 2°C to 8°C Flash Point: 50°(122°F) Refractive Index: 1.5125 Odour: Pungent, makes eyes water Boiling Point: 56-58°/10mm Beilstein Reference: 1210127 UN No.: 3080 MDL No.: MFCD00001996

Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 56…58 °C (10 mmHg)
Density: 1.221
Flash Pt: 50 °C (122 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001996
CAS Number: 16744-98-2
EINECS: 240-804-0
UN: 3080
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR