Cadmium(II) iodide 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cadmium iodide

A18079.14 A18079.22
A18079.14EA 38.2 EUR
A18079.14 A18079.22
Cadmium(II) iodide 98
Cadmium (II) iodide

Purity: 98% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Odour: Odourless Melting Point: 387-389° Boiling Point: 787° Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920

Formula: CdI₂
MW: 366.22 g/mol
Boiling Pt: 787 °C
Melting Pt: 387…389 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00010920
CAS Number: 7790-80-9
EINECS: 232-223-6
UN: 2570
Merck Index: 12,01657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR