Carboxymethyl cellulose sodium salt

Supplier: Thermo Fisher Scientific

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

A18105.0E A18105.36
A18105.0EEA 254 EUR
A18105.0E A18105.36
Carboxymethyl cellulose sodium salt
Carboxymethyl cellulose sodium salt

CAS No.: 9004-32-4 Melting Point: 300° Merck Reference: 14,1829 MDL No.: MFCD00081472

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 14,1829

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR