Styrene ≥99% stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Phenylethene, Phenylethylene, Vinylbenzene

Stabilization: 10-15 ppm Stabilised with 4-tert-butylpyrocatechol (TBC)

A18481.0F A18481.AE A18481.AP
A18481.0FEA 71.6 EUR
A18481.0F A18481.AE A18481.AP
Styrene ≥99% stabilised
Styrene

Stabilized with 10-15 ppm 4-tert-butylcatechol


Purity: 99% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Refractive Index: 1.5460 Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612

Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 145…146 °C
Melting Pt: –31 °C
Density: 0.906
Flash Pt: 31 °C (88 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008612
CAS Number: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3 III
Merck Index: 14,8860

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR