Sodium nitrite 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Diazotizing salts

A18668.0I A18668.36 A18668.0B
A18668.0IEA 162 EUR
A18668.0I A18668.36 A18668.0B
Sodium nitrite 98
Sodium nitrite

Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118

Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320
Melting Pt: 271 °C (dec.)
Density: 2.17
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1 III
Merck Index: 13,08721

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR