Methyl-5-bromovalerate 97

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 5-Bromovalerate, 5-Bromovaleric acid methyl ester

A19072.06 A19072.14 A19072.22
A19072.06EA 54.4 EUR
A19072.06 A19072.14 A19072.22
Methyl-5-bromovalerate 97
Methyl 5-bromovalerate

Purity: 97% CAS No.: 5454-83-1 Molecular Formula: Br(CH2)4CO2CH3 Formula Weight: 195.06 Flash Point: 99°(210°F) Refractive Index: 1.4630 Boiling Point: 79-80°/4mm Beilstein Reference: 1749649 MDL No.: MFCD00000265

Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06 g/mol
Boiling Pt: 79…80 °C (4 mmHg)
Density: 1.363
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000265
CAS Number: 5454-83-1
EINECS: 226-709-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR