Tris(perfluorobutyl)amine tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

A19712.06 A19712.14
A19712.06EA 19.6 EUR
A19712.06 A19712.14
Tris(perfluorobutyl)amine tech. 90%, Technical Grade
Tris(perfluorobutyl)amine

Purity: 90% CAS No.: 311-89-7 Molecular Formula: [CF3(CF2)3]3N Formula Weight: 671.10 Refractive Index: 1.2890 Melting Point: -52° Boiling Point: 177-180° Beilstein Reference: 1813883 MDL No.: MFCD00000436

Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 177…180 °C
Melting Pt: –52 °C
Density: 1.880
Storage Temperature: Ambient
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR