Tris-Acetate-EDTA (TAE) buffer, 10X, molecular biology grade
Supplier: PanReac AppliChem
A4227.1000 A4227.5000
A4227.1000EA 78 EUR
A4227.1000 A4227.5000
Tris-Acetate-EDTA (TAE) buffer, 10X, molecular biology grade
Buffers Electrophoresis Buffers Running Buffers
10X concentrated solution of 40 mM Tris, 20 mM glacial acetic acid and 1 mM EDTA.Na₂, pH 8,5 ±0,2.
DNases, RNases, and proteases not detectable.