Proteinase K solution

Supplier: PanReac AppliChem

A4392.0001 A4392.0005 A4392.0010
A4392.0001EA 44.8 EUR
A4392.0001 A4392.0005 A4392.0010
Proteinase K solution
Enzymes

Proteinase K is used to destruct proteins in cell lysates (tissue, cultured cells) and to liberate nucleic acids, since it very effectively digests DNases and RNases.


  • DNases/RNases not detectable
  • Proteinase K min. 600 mAnsonU/ml
  • GHS symbols: GHS08
  • GHS signal word: Danger


Composition:
CaCl₂ · 2H₂O 1 mM
Glycerol (v/v) 40%
Proteinase K 20 mg/ml
Tris · HCl (pH 7,5) 10 mM

Order Now

SPECIFICATIONS

Enzyme name Proteinase K solution
Environmentally Preferable
Enzyme synonyms Protease K solution 20 mg/ml, Endopeptidase K solution 20 mg/ml, Tritirachium alkaline proteinase solution 20 mg/ml, Tritirachium album serine proteinase solution 20 mg/ml, Peptidase K solution 20 mg/ml
Source
Species
Concentration 20 mg/ml
Storage conditions 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR