VWR®, Pre-weighed dehydrated culture media in sachets
Supplier: VWR Chemicals
S246SAC5EA
20.8
EUR
S246SAC5
A887SAC5
S378SAC5
B202SAC5
B455SAC5
A990SAC5
A935SAC5
B453SAC5
A484SAC5
A265SAC5
A919SAC5
A857SAC5
B857SAC5
A865SAC5
B923SAC5
A704SAC5
A979SAC5
S821SAC5
B710SAC5
A389SAC5
A971SAC5
A882SAC5
A355SAC5
A925SAC5
A188SAC5
A890SAC5
A869SAC5
B700SAC5
A807SAC5
A836SAC5
A975SAC5
B475SAC5
A128SAC5
A875SAC5
B989SAC5
B982SAC5
A986SAC5
A760SAC5
B206SAC5
B254SAC5
A487SAC5
B872SAC5
A333SAC5
A751SAC5
A961SAC5
B419SAC5
A794SAC5
B901SAC5
A438SAC5
A902SAC5
A700SAC5
A852SAC5
A993SAC5
A940SAC5
A391SAC5
B397SAC5
A167SAC5
B167SAC5
B418SAC5
A415SAC5
B415SAC5
A851SAC5
A427SAC5
A731SAC5
B811SAC5
B833SAC5
B336SAC5
A540SAC5
A723SAC5
A338SAC5
B739SAC5
A336SAC5
A870SAC5
B878SAC5
A317SAC5
B317SAC5
A318SAC5
B318SAC5
B117SAC5
B734SAC5
A315SAC5
S834SAC5
A322SAC5
A817SAC5
A329SAC5
B322SAC5
A253SAC5
A259SAC5
A261SAC5
B261SAC5
S158SAC5
A729SAC5
B150SAC5
A383SAC5
A234SAC5
S460SAC5
A382SAC5
A407SAC5
A193SAC5
A388SAC5
B387SAC5
A387SAC5
A457SAC5
A777SAC5
A912SAC5
A174SAC5
A797SAC5
B706SAC5
B894SAC5
B884SAC5
S358SAC5
A750SAC5
S272SAC5
A830SAC5
A785SAC5
B368SAC5
S365SAC5
A304SAC5
A709SAC5
A748SAC5
A499SAC5
B499SAC5
B396SAC5
A483SAC5
B494SAC5
A491SAC5
A190SAC5
A498SAC5
B495SAC5
A433SAC5
A774SAC5
VWR®, Pre-weighed dehydrated culture media in sachets
Media for Microbiology
Dehydrated Media for Microbiology
Dehydrated culture media in small sachets ready to make up exactly 500 ml of medium.
- Quick: Simply open the sachet and you have the right quantity of powder to prepare 500 ml of medium
- Safe: No more need to transfer and weigh powder
- Always fresh media for best quality: All the dehydrated media inside the sachet is always used immediately after opening the packaging
- Space saving: You can have tens of dehydrated media stored in a very small space inside your laboratory
- Saving money: You buy only the quantity of dehydrated media that you really need
Every pack contains 5 sachets each of which allows to prepare 500 ml of medium.
Informace pro objednávání: About 140 different culture media available.