iFluor® 647 fluorescent probe

Supplier: AAT BIOQUEST
Warning

1019
AATB1019EA 292 EUR
AATB1019
iFluor® 647 fluorescent probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b51f69dac7c33ecbaa2b6678b81ffcd609259fc&vid=2376eacde46479760c657f6682a00015b4e582b1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR