iFluor® 488, Reactive Form_Succinimidyl Ester
Supplier: AAT BIOQUEST
71023 71503 71553 1023
AATB71023EA 114 EUR
AATB71023 AATB71503 AATB71553 AATB1023
iFluor® 488, Reactive Form_Succinimidyl Ester
iFluor®
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Upozornení: For research use only.