Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PE (Phycoerythrin)/Texas Red®)

Supplier: AAT BIOQUEST
103401S0 103401S1 103401S2
AATB103401S0EA 319 EUR
AATB103401S0 AATB103401S1 AATB103401S2
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PE (Phycoerythrin)/Texas Red®)
Antibodies
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR