D(-)-Luciferin potassium salt 95

Supplier: AAT BIOQUEST
Warning

Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate

12505 12506 12507
AATB12505EA 120 EUR
AATB12505 AATB12506 AATB12507
D(-)-Luciferin potassium salt 95
D(-)-Luciferin potassium salt

Luciferin is the most popular and versatile bioluminescent substrate.

Formula: C₁₁H₇KN₂O₃S₂
MW: 318.41 g/mol
MDL Number: MFCD00044928
CAS Number: 115144-35-9
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b95864949694c304bf77dfbead0e719455e8f25b&vid=77c89f90cee052e068adbbe5db6ae96089d6dfbe&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR