Amplite® Fluorimetric Total NAD and NADH Assay Kit *Red Fluorescence*
Supplier: AAT BIOQUEST
15257
AATB15257EA 460 EUR
AATB15257
Amplite® Fluorimetric Total NAD and NADH Assay Kit *Red Fluorescence*
Assays Enzyme Assays
Nicotinamide adenine dinucleotide (NAD+) and nicotinamide adenine dinucleotide phosphate (NADP+) are two important cofactors found in cells.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Upozornení: For research use only.