Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Supplier: AAT BIOQUEST
17630
AATB17630EA 281 EUR
AATB17630
Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Assays Nucleic Acid Assays
Helixyte™ Green fluorimetric total nucleic acid quantitation kit is optimised for measuring the total amounts of nucleic acids with a fluorescence microplate reader.
  • Optimised for microplate readers
  • Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green all reagent, dilution buffer, and pre-diluted DNA standards..
Balení: Plastic vial.
Upozornení: For research use only.