Mag-Fluo-4 potassium salt

Supplier: AAT BIOQUEST
Warning

20400
AATB20400EA 230 EUR
AATB20400
Mag-Fluo-4 potassium salt
Mag-Fluo-4 potassium salt

The cell-impermeant Mag-Fluo-4 is an analog of Fluo-4 with a Kd for Mg ion of 4.7 mM and a Kd for Ca ion of 22 µM, making it useful as an intracellular Mg ion indicator as well as a low-affinity Ca ion indicator.


  • Suitable for either mg or ca detection
  • Steady signal
  • Cell impermeant


Upozornení: For research use only.

MW: 681.77 g/mol
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR