Rhod-2 trisodium salt

Supplier: AAT BIOQUEST
Warning

21068
AATB21068EA 307 EUR
AATB21068
Rhod-2 trisodium salt
Rhod-2 trisodium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Long-wavelength emission
  • Cell impermeant


Upozornení: For research use only.

Formula: C₄₀H₃₉N₄Na₃O₁₁
MW: 945 g/mol
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR