Rhod-4™ azide
Supplier: AAT BIOQUEST
21125
AATB21125EA 687 EUR
AATB21125
Rhod-4™ azide
Rhod-4™ azide
Rhod-4™ azide is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.
  • Stable for 12 months
Quality tested in-house
Upozornení: For Research Use Only