iFluor® 647, Styramide *Superior Replacement for Alexa Fluor 647 tyramide*
Supplier: AAT BIOQUEST
45045
AATB45045EA 460 EUR
AATB45045
iFluor® 647, Styramide *Superior Replacement for Alexa Fluor 647 tyramide*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
  • 10 to 50X higher fluorescence
  • Improved precision and sensitivity
  • More efficient use of primary antibody
  • Enhanced water solubility
  • More photostable
Upozornení: For research use only.