ReadiLink™ Rapid XFD647 Antibody Labelling Kit
Supplier: AAT BIOQUEST
5740
AATB5740EA 687 EUR
AATB5740
ReadiLink™ Rapid XFD647 Antibody Labelling Kit
Protein Linking and Labeling Systems
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped at RT.
Upozornení: Research use only.