iFluor® 750, Reactive Form_Succinimidyl Ester
Supplier: AAT BIOQUEST
1037 71037 71515 71565
AATB1037EA 307 EUR
AATB1037 AATB71037 AATB71515 AATB71565
iFluor® 750, Reactive Form_Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are developed for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Upozornení: For research use only.