iFluor® 800, Reactive Form_Succinimidyl Ester
Supplier: AAT BIOQUEST
1379 71379 71525 71575
AATB1379EA 460 EUR
AATB1379 AATB71379 AATB71525 AATB71575
iFluor® 800, Reactive Form_Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright fluorescence
  • High photostability
  • Low quenching on proteins
  • Useful for in-vivo imaging
Upozornení: For research use only.