iFluor® 860, Reactive Form_Succinimidyl Ester
Supplier: AAT BIOQUEST
1409 71409 71529 71579
AATB1409EA 460 EUR
AATB1409 AATB71409 AATB71529 AATB71579
iFluor® 860, Reactive Form_Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright fluorescence
  • High photostability
  • Low quenching on proteins
  • Useful for in-vivo imaging
Upozornení: For research use only.