Anti-CD3 Rabbit Polyclonal Antibody

Supplier: Abcam
AB5690-100
ABCAAB5690-100EA 974 EUR
ABCAAB5690-100
Anti-CD3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR