Anti-IgG Goat Polyclonal Antibody (Cy5®)

Supplier: Abcam
AB6565-500
ABCAAB6565-500EA 471 EUR
ABCAAB6565-500
Anti-IgG Goat Polyclonal Antibody (Cy5®)
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR