Anti-HSPB1 Rabbit Polyclonal Antibody

Supplier: Abcam
AB90538-50
ABCAAB90538-50EA 918 EUR
ABCAAB90538-50
Anti-HSPB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR