Anti-IHH Mouse Monoclonal Antibody

Supplier: Abgent
AM1981A
ABGEAM1981AEA 354 EUR
ABGEAM1981A
Anti-IHH Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: IHH (indian hedgehog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol IHH
Antigen name Indian Hedgehog Homolog
Environmentally Preferable
Antigen synonyms BDA1|HHG-2|HHG2
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 363CT4.1.6
Reactivity Human
Host Mouse
Gene ID 3549
Isotype IgG3
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope C-Terminal
Immunogen This IHH Monoclonal antibody is generated from mouses immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human IHH.
Sequence SLISSENFPELKVIEMC
Form Ascites
Molecular weight 45.251 kDa
Cross adsorption No
Storage buffer PBS with 0,09% (W/V) sodium azide, pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR