Anti-HDAC4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1104A
ABGEAP1104AEA 354 EUR
ABGEAP1104A
Anti-HDAC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDAC4 (histone deacetylase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR