Anti-PDPN Rabbit Polyclonal Antibody

Supplier: Abgent
AP2016B
ABGEAP2016BEA 354 EUR
ABGEAP2016B
Anti-PDPN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDPN (podoplanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol PDPN
Antigen name Podoplanin
Environmentally Preferable
Antigen synonyms T1A-2|AGGRUS|T1A|HT1A-1|GP36|Gp38|PA2.26|T1A2|TI1A|OTS8|GP40
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Epitope N-Terminal ( 48–63 aa)
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR