Anti-USP28 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2152A
ABGEAP2152AEA 354 EUR
ABGEAP2152A
Anti-USP28 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR