(4-Aminophenyl)acetonitrile 99%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide

103210100
ACRO103210100EA 89.8 EUR
ACRO103210100
(4-Aminophenyl)acetonitrile 99%
(4-Aminophenyl)acetonitrile
Formula: C₈H₈N₂
MW: 132.17 g/mol
Melting Pt: 42…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007912
CAS Number: 3544-25-0
EINECS: 222-587-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 41 to 46 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR